Learn More
Branson Ultrasonics™ Sonifier™ Replacement Flat Tip For Tapped Horn
Used with 1/2 in. (12mm) tapped horn for general processing
Brand: Branson Ultrasonics™ 101-148-013
Additional Details : Weight : 0.99000kg
Description
Used with 1/2 in. (12mm) tapped horn for general processing