missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26004-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Negatively regulates MAP kinase activation by limiting the formation of Raf/MEK complexes probably by inactivation of the KSR1 scaffold protein. Also acts as a Ras responsive E3 ubiquitin ligase that, on activation of Ras, is modified by auto-polyubiquitination resulting in the release of inhibition of Raf/MEK complex formation. May also act as a cytoplasmic retention protein with a role in regulating nuclear transport.Specifications
BRAP | |
Polyclonal | |
Unconjugated | |
BRAP | |
BRAP, BRAP2, BRCA1 associated protein, IMP, RING finger protein 52, RNF52 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8315 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z569 | |
BRAP | |
BRAP Fusion Protein Ag23279 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |