Läs mer
Tocris Bioscience™ BRD 73954
GREENER_CHOICE
Produktkod. 18790528
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18790528 10 mg
18765798 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18790528 Leverantör Tocris Bioscience™ Leverantörsnummer 6284/10
Dual histone deacetylase (HDAC) 6/8 inhibitor
BRD 73954 is a dual histone deacetylase (HDAC) 6/8 inhibitor. IC50 values are 0.036 μM, 0.12 μM, 9 μM, >33 μM, for HDAC6, HDAC8, HDAC2 and HDAC4, respectively.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N1-Hydroxy-N3-(2-phenylethyl)-1,3-benzenedicarboxamide
CAS 1440209-96-0
Analysprocentintervall >98%
Kvantitet 10 mg
Molekylformel C16H16N2O3
Formel vikt Observed MW: 284.31