Läs mer
BRD73954, MedChemExpress
Produktkod. 16493607
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
10mg
25mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16493607 50 mg -
16450068 5 mg -
16407598 10 mg 10mg
16403616 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16493607 Leverantör Medchem Express Leverantörsnummer HY18700
MedChemExpress BRD73954 ia a potent and selective HDAC inhibitor with IC50 of 36 nM and 120 nM for HDAC6 and HDAC8, respectively.
BRD73954 ia a potent and selective HDAC inhibitor with IC50 of 36 nM and 120 nM for HDAC6 and HDAC8, respectively.
Specifikationer
Kemiskt namn eller material BRD73954
Fysisk form Solid
CAS 1440209-96-0
Färg Off-White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C16H16N2O3
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 25 mg/mL (87.93 mM; Need ultrasonic)
LEDER O=C(C1=CC=CC(C(NCCC2=CC=CC=C2)=O)=C1)NO
Molekylvikt (g/mol) 284.31
Formel vikt 284.31
Procent renhet 98.0%