Learn More
BRD8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10476-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16826883
  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021
Description
Description
The protein encoded by this gene interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. This protein contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced transcript variants that encode distinct isoforms have been identified.
Specifications
Specifications
BRD8
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H0E9
BRD8, bromodomain containing 8, p120, SMAP, SMAP2, TrCP120
Rabbit
IgG
20 μL
-20°C
Primary
10902
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence
Unconjugated
BRD8
Liquid
BRD8
BRD8 Fusion Protein Ag0790
Antigen Affinity Chromatography