Learn More
Invitrogen™ eBioscience™ BrdU Kit for IHC/ICC Immunofluorescence eFluor™ 660
Contains reagents and buffers to identify and examine proliferating cells by immunohistochemical or immunocytochemical analysis. Optimized for IHC with both frozen and formalin-fixed paraffin-embedded BrdU-labeled mouse intestine and ICC of BrdU-pulsed HeLa cells grown on culture slides.
Brand: Invitrogen™ 8850-6599-45
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide and formaldehyde
Includes: BrdU Fixation Buffer
BrdU Antigen Retrieval Solution (10X)
BrdU Blocking Solution
Anti-BrdU Biotin (clone BU20A) : 10 μg
Streptavidin for BrdU IHC/ICC eFluor 660 : 25 μg
Description
Cells are labeled in vitro or in vivo with 5-bromo-2’-deoxyuridine (BrdU), a synthetic analog of thymidine, which is incorporated into DNA in place of thymidine during the S-phase of the cell cycle. Following fixation and antigen retrieval steps, cells or tissue sections are stained for BrdU incorporation and visualized using a streptavidin-conjugated fluorophore.Specifications
BrdU | |
eBioscience™ | |
IHC/ICC BrdU Kit |
Fluorometric | |
ICC (Immunochemistry), IHC (Immunohistochemistry) | |
All species |
For Research Use Only. Not for use in diagnostic procedures.