Learn More
Grant Instruments™ Bridge Plate
Bridge plate fits T Optima heating circulator models to Y6 & W6 tanks
Brand: Grant Instruments™ T-BYWS
Additional Details : Weight : 4.00000kg
Description
- Bridge plate fits T Optima heating circulator models to Y6 & W6 tanks
Specifications
Y6 and W6 Tanks | |
Bridge Plate | |
312 x 216 x 168 mm |