missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Crocein, Thermo Scientific Chemicals

Catalog Number p-7022049
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
5413-75-2
C22H16N4Na2O7S2
558.491
MFCD00003907
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
C.I. 27290; Crocein Scarlet MOO
88641610
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
This item is not returnable. View return policy
5413-75-2
C22H16N4Na2O7S2
558.491
MFCD00003907
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
C.I. 27290; Crocein Scarlet MOO
88641610
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Solubility Information Soluble in water
Formula Weight 556.49
Quantity 50 g
Chemical Name or Material Brilliant Crocein

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'