missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRIP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529889
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LIN54 is a component of the LIN, or DREAM, complex, an essential regulator of cell cycle genes.Specifications
BRIP1 | |
Polyclonal | |
Unconjugated | |
BRIP1 | |
3110009N10Rik; 8030460J03Rik; ATP-dependent RNA helicase BRIP1; BACH 1; Bach1; BACH1 BRIP1; BRCA1 interacting protein C-terminal helicase 1; BRCA1/BRCA2-associated helicase 1; BRCA1-associated C-terminal helicase 1; BRCA1-binding helicase-like protein BACH1; BRCA1-interacting protein 1; BRCA1-interacting protein C-terminal helicase 1; BRIP 1; Brip1; FACJ; FANCJ; FANCJ protein; Fanconi anemia group J protein; Fanconi anemia group J protein homolog; FLJ90232; MGC126521; MGC126523; OF; Protein FACJ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83990 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BX63 | |
BRIP1 | |
Recombinant fragment corresponding to a region within amino acids 997 and 1221 of Human FANCJ | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |