Läs mer
Tocris Bioscience™ BRL 50481
Produktkod. 18024942
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18024942 10 mg
18093962 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18024942 Leverantör Tocris Bioscience™ Leverantörsnummer 2237/10
Selective PDE7 inhibitor
Selective, substrate-competitive inhibitor of phosphodiesterase (PDE) 7 (Ki = 180 nM). Displays > 200-fold selectivity over PDE1B, PDE1C, PDE2, PDE3, PDE4A4 and PDE5.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 433695-36-4
Molekylformel C9H12N2O4S
Specifikationer
Kemiskt namn eller material BRL 50481
Kvantitet 10 mg
Formel vikt 244.26
Renhet >99%
Molekylvikt (g/mol) 244.265