Läs mer
Brobactam Sodium Salt, TRC
Produktkod. 30329363
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30329363 10 mg
1 options
Produktkod. 30329363 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB67785010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Brobactam Sodium Salt
CAS 73335-78-1
Molekylformel C8H9BrNNaO3S
Kvantitet 10 mg
Synonym (2S,5R,6R)-6-Bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Sodium Salt, Sulbactam EP Impurity D Sodium Salt, [2S-(2α,5α,6β)]-6-Bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Sodium Salt, 6β-Bromopenicillanic Acid Sodium Salt, BRL 25214 Sodium Salt
InChI formel InChI=1S/C8H10BrNO3S.Na/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8;/h3-4,6H,1-2H3,(H,12,13);/q;+1/p-1/t3-,4+,6-;/m1./s1
LEDER [H][C@]12N([C@@H](C(O[Na])=O)C(C)(C)S2)C([C@H]1Br)=O
IUPAC-namn sodium;(2S,5R,6R)-6-bromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molekylvikt (g/mol) 302.12