Läs mer
Brocresine Hydrochloride, LoGiCal
Produktkod. 30459682
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30459682 5 mg
1 options
Produktkod. 30459682 Leverantör LoGiCal Leverantörsnummer LGCFOR1839.00
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Brocresine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Brocresine Hydrochloride
CAS 91468-63-2
Analyt- eller komponentnamn Brocresine Hydrochloride
Frakt skick Room Temperature
Molekylformel C7 H8 Br N O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Phenol, 5-[(aminooxy)methyl]-2-bromo-, hydrochloride (1:1), Benzyloxyamine, 4-bromo-3-hydroxy-, hydrochloride (7 CI)
InChI formel InChI=1 S/C7H8BrNO2.ClH/c8-6-2-1-5(4-11-9)3-7(6)10;/h1-3,10 H,4,9H2;1 H
LEDER Cl.NOCc1ccc(Br)c(O)c1
IUPAC-namn 5-(aminooxymethyl)-2-bromophenol;hydrochloride
Industry Type Forensics
Molekylvikt (g/mol) 254.51
Kvantitet 5 mg
Formel vikt 252.9505 g/mol