Description
- Bromelain may be used to inhibit the biosysnthesis of proinflammatory prostaglandins
- It may be used to reduce clotting efficiency
- Bromelain, from pineapple stem, has been used to make enzymatic hydrolysates of honeybee-collected pollen
Specifications
Specifications
| CAS | 9001-00-7 |
| Color | Tan |
| MDL Number | MFCD00081423 |
| Quantity | 100 g |
| Solubility Information | Readily soluble in water, insoluble in most organic solvents such as acetone, ether, ethanol and methanol. |
| Formula Weight | Approximately 33,000. (Lit.) |
| Grade | Not Available |
| Physical Form | Powder |
| Chemical Name or Material | BROMELAIN from Pineapple |