missing translation for 'onlineSavingsMsg'
Learn More

Bromindione, MedChemExpress

Product Code. 16444507
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Kvantitet:
50 mg
This item is not returnable. View return policy

Product Code. 16444507

missing translation for 'mfr': Medchem Express HYB091750mg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

MedChemExpress Bromindione is a potent, long-acting, inandione-derived, oral anticoagulant compound.

Bromindione is a potent, long-acting, inandione-derived, oral anticoagulant compound.

Specifications

Kemiskt namn eller material Bromindione
CAS 1146-98-1
Färg Brown
Anteckningar om renhetsgrad Research
Molekylformel C15H9BrO2
Kvantitet 50 mg
Synonym Fluidane Halinone
Löslighetsinformation DMSO : 250 mg/mL (830.21 mM; Need ultrasonic)
LEDER O=C1C(C2=CC=C(Br)C=C2)C(C3=C1C=CC=C3)=O
Molekylvikt (g/mol) 301.13
Formel vikt 301.13
Procent renhet 98.02%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.