Läs mer
Bromkal 73-5PE, TRC
Produktkod. 17948904
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17948904 250 mg
17947769 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17948904 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB678800250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35109-60-5
Molekylformel C9 H7 Br5 O
Molekylvikt (g/mol) 530.67
Synonym 1,3,5-Tribromo-2-(2,3-dibromopropoxy)benzene, 2,3-Dibromopropyl 2,4,6-Tribromophenyl Ether, 2,4,6-Tribromophenyl 2,3-Dibromopropyl Ether
IUPAC-namn 1,3,5-tribromo-2-(2,3-dibromopropoxy)benzene
LEDER BrCC(Br)COc1c(Br)cc(Br)cc1Br
Specifikationer
Kemiskt namn eller material Bromkal 73-5PE
Kvantitet 250 mg
InChI formel InChI=1S/C9H7Br5O/c10-3-6(12)4-15-9-7(13)1-5(11)2-8(9)14/h1-2,6H,3-4H2
Formel vikt 525.6414