Läs mer
Bromo 2,4,6-Tri-O-acetyl-3-O-(2,3,4,6-tetra-O-acetyl -b-D-glucopyranosyl)-Alpha-D-glucopyranoside, TRC
Produktkod. 30342810
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30342810 5 mg
1 options
Produktkod. 30342810 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6883355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bromo 2,4,6-Tri-O-acetyl-3-O-(2,3,4,6-tetra-O-acetyl -b-D-glucopyranosyl)-alpha-D-glucopyranoside
CAS 23202-66-6
Molekylformel C26H35BrO17
Kvantitet 5 mg
Synonym 3-O-(2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl)-alpha-D-glucopyranosyl Bromide Triacetate, Acetobromolaminaribiose
InChI formel InChI=1S/C26H35BrO17/c1-10(28)35-8-17-19(37-12(3)30)21(23(25(27)42-17)40-15(6)33)44-26-24(41-16(7)34)22(39-14(5)32)20(38-13(4)31)18(43-26)9-36-11(2)29/h17-26H,8-9H2,1-7H3/t17-,18-,19-,20-,21+,22+,23-,24-,25+,26+/m1/s1
LEDER CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(=O)C)[C@@H](O[C@@H]2O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]2OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-3,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-4-yl]oxyoxan-2-yl]methyl acetate
Molekylvikt (g/mol) 699.45
Formel vikt 698.1058