missing translation for 'onlineSavingsMsg'
Learn More

α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-77139
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17201-43-3
C8H6BrN
196.05
MFCD00001829
UMLFTCYAQPPZER-UHFFFAOYSA-N
4-cyanobenzyl bromide, 4-bromomethyl benzonitrile, p-cyanobenzyl bromide, alpha-bromo-p-tolunitrile, benzonitrile, 4-bromomethyl, 4-bromomethyl-benzonitrile, alpha-bromo-p-toluonitrile, p-bromomethyl benzonitrile, 4-cyano benzyl bromide, benzonitrile, p-bromomethyl
86996
4-(bromomethyl)benzonitrile
C1=CC(=CC=C1CBr)C#N
This item is not returnable. View return policy
17201-43-3
C8H6BrN
196.05
MFCD00001829
UMLFTCYAQPPZER-UHFFFAOYSA-N
4-cyanobenzyl bromide, 4-bromomethyl benzonitrile, p-cyanobenzyl bromide, alpha-bromo-p-tolunitrile, benzonitrile, 4-bromomethyl, 4-bromomethyl-benzonitrile, alpha-bromo-p-toluonitrile, p-bromomethyl benzonitrile, 4-cyano benzyl bromide, benzonitrile, p-bromomethyl
86996
4-(bromomethyl)benzonitrile
C1=CC(=CC=C1CBr)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 113°C to 117°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CN
Quantity 10 g
Beilstein 09, 499
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in inert organic solvents
Formula Weight 196.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α-Bromo-p-tolunitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • a-Bromo-p-tolunitrile
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'