Läs mer
Bromobimane, MedChemExpress
Produktkod. 16402526
Encompass_Preferred
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16402526 25 mg
16422768 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16402526 Leverantör Medchem Express Leverantörsnummer HY100041
MedChemExpress Bromobimane is essentially nonfluorescent and converts into fluorescent products when reacts with small thiols.
Bromobimane is essentially nonfluorescent and converts into fluorescent products when reacts with small thiols.
Specifikationer
Kemiskt namn eller material Bromobimane
CAS 71418-44-5
Anteckningar om renhetsgrad Research
Molekylformel C10H11BrN2O2
Synonym Monobromobimane
Löslighetsinformation DMSO : ≥ 86.6 mg/mL (319.43 mM)
LEDER O=C1N(C(C(C)=C2C)=O)N2C(CBr)=C1C
Molekylvikt (g/mol) 271.11
Formel vikt 271.11
Procent renhet 98.0%
Kvalitet Research
Färg Yellow
Kvantitet 25 mg
Fysisk form Solid