Läs mer
Bromocresol Green, Pure, Indicator Grade, Fisher Chemical™
Produktkod. 10355170 Handla allt Fisher Chemical Produkter
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
10355170 1 g 1g
10223900 5 g 5g
10263180 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 10355170 Leverantör Fisher Chemical Leverantörsnummer B/4320/43
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Specifikationer
Smältpunkt 225°C
CAS 76-60-8
Molekylformel C21H14Br4O5S
MDL-nummer 5874
Formel vikt 698.01g/mol
Förpackning Glass Jar
Färg Beige
Kvantitet 1 g
Fysisk form Powder