Läs mer
Bromometylmetanetiosulfonat, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Bromomethyl Methanethiosulfonate |
| CAS | 887353-78-8 |
| Molekylformel | C2 H5 Br O2 S2 |
| Kvantitet | 100 mg |
| Synonym | Methanesulfonothioic acid, S-(bromomethyl) ester, Bromo-methylsulfonylsulfanylmethane, Bromomethyl methanethiosulfonate, Methanesulfonothioic acid S-(bromomethyl) ester |
| InChI formel | InChI=1S/C2H5BrO2S2/c1-7(4,5)6-2-3/h2H2,1H3 |
| LEDER | CS(=O)(=O)SCBr |
| IUPAC-namn | Brom(metylsulfonylsulfanyl)metan |
| Molekylvikt (g/mol) | 205.09 |
| Formel vikt | 203.8914 |