Läs mer
Bromopentacarbonylrhenium(I), TRC
Produktkod. 30343241
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30343241 500 mg
1 options
Produktkod. 30343241 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB740015500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bromopentacarbonylrhenium(I)
CAS 14220-21-4
Molekylformel C5BrO5Re
Kvantitet 500 mg
Synonym Bromopentacarbonylrhenium, Bromopentacarbonylrhenium(I)
InChI formel InChI=1S/5CO.BrH.Re/c5*1-2;;/h;;;;;1H;/q;;;;;;+1/p-1
LEDER [Re]Br.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
IUPAC-namn bromorhenium;carbon monoxide
Molekylvikt (g/mol) 406.16
Formel vikt 405.85