Läs mer
Bromopropylate, TRC
Produktkod. 30348504
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30348504 250 mg
1 options
Produktkod. 30348504 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB687745250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bromopropylate
CAS 18181-80-1
Molekylformel C17 H16 Br2 O3
Kvantitet 250 mg
Synonym Benzilic acid, 4,4'-dibromo-, isopropyl ester (8CI), Acarol, Akpin, Akpinol, Bromopropylate, Folbex VA, GS 19851, Geigy 19851, Isopropyl 4,4'-dibromobenzilate, Isopropyl bis(4-bromophenyl)glycolate, Isopropyl dibromobenzilate, Nanocron, Neoron, Neoron (pesticide), Phenisobromolate
InChI formel InChI=1S/C17H16Br2O3/c1-11(2)22-16(20)17(21,12-3-7-14(18)8-4-12)13-5-9-15(19)10-6-13/h3-11,21H,1-2H3
LEDER CC(C)OC(=O)C(O)(c1ccc(Br)cc1)c2ccc(Br)cc2
IUPAC-namn propan-2-yl 2,2-bis(4-bromophenyl)-2-hydroxyacetate
Molekylvikt (g/mol) 428.12
Formel vikt 425.9466