missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BromoTag Antibody, Alexa Fluor™ 405, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP3-17999AF405
This item is not returnable.
View return policy
Description
BromoTag Polyclonal antibody specifically detects BromoTag in Nonspecies specific samples. It is validated for Western BlotSpecifications
BromoTag | |
Polyclonal | |
Western Blot | |
Brd4 bromodomain L387A variant, Brd4BD2L387A, BromoTag degron | |
Partial recombinant protein made to the second bromodomain of Brd4 with a L387A mutation (see Bond, et al PMID 34652918). | |
0.1 mL | |
Inhibitors Activators and Regulators, Ubiquitin Proteasome Pathway | |
Non-species specific | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Sheep | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4C in the dark. |