missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Sultone Form, ACS Reagent, Reag. Ph. Eur., Honeywell Fluka™

Catalog Number 15633840
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
76-59-5
C27H28Br2O5S
624.384
MFCD00005872
NUHCTOLBWMJMLX-UHFFFAOYSA-N
3′,3″-Dibromothymolsulfonphthalein
6450
CHEBI:86155
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
This item is not returnable. View return policy
76-59-5
C27H28Br2O5S
624.384
MFCD00005872
NUHCTOLBWMJMLX-UHFFFAOYSA-N
3′,3″-Dibromothymolsulfonphthalein
6450
CHEBI:86155
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Indicator, ACS Reagent, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
UN Number NONH for all modes of transport
Beilstein 373934
Formula Weight 624.38g/mol
Grade ACS Reagent
Packaging Glass Bottle
Suitability Complies for clarity of solution
Visual Transition Interval 5.8 to 7.4
Color Violet
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Bromothymol Blue Sultone Form, Reag. Ph. Eur.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'