Läs mer
Tocris Bioscience™ Bropirimine
GREENER_CHOICE
Produktkod. 18084852
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18084852 50 mg
18033812 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18084852 Leverantör Tocris Bioscience™ Leverantörsnummer 4316/50
Immunomodulatory and antitumor compound
Immunomodulatory and antitumor compound. Induces interferon (IFN)-α/β production; augments natural killer (NK) cell activity. Orally active.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56741-95-8
Molekylformel C10H8BrN3O
Specifikationer
Kemiskt namn eller material Bropirimine
Kvantitet 50 mg
Formel vikt 266.09
Renhet >99%
Molekylvikt (g/mol) 266.098