Learn More
Hirschmann™ Amber Glass Stoppers
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
Brand: Hirschmann™ 8902370
Additional Details : Weight : 0.25000kg
Description
- Acc. to DIN 12 252
Specifications
Stopper | |
Glass | |
NS 14/23 |
1/Ea. | |
For tight sealing of standard ground vessels |