missing translation for 'onlineSavingsMsg'
Läs mer

(-)-Brucine, Sigma-Aldrich™

Produktkod. 15814401
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 g
Förpackningsstorlek:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15814401 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15814401 Leverantör Sigma Aldrich Leverantörsnummer 8183080050
 Begränsad artikel
Beräknad leverans: 28-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For resolution of racemates for synthesis

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 357-57-3
Smältpunkt 175°C to 178°C
MDL-nummer MFCD00005942
Kvantitet 50 g
Synonym (-)-Brucine, 2,3-Dimethoxy strychnine, (-)-Brucine
Molekylvikt (g/mol) 394.46
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.