missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRUNOL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19078-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
RNA-binding protein implicated in the regulation of pre-mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of TNNT2 in a muscle-specific splicing enhancer (MSE)-dependent manner. Promotes also exon exclusion of INSR pre-mRNA.Specifications
BRUNOL6 | |
Polyclonal | |
Unconjugated | |
CELF6 | |
Bruno like protein 6, BRUNOL6, CELF 6, CELF6, RNA binding protein BRUNOL 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
300758, 60677, 76183 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TN33, Q96J87 | |
CELF6 | |
BRUNOL6 Fusion Protein Ag4970 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |