Läs mer
Tocris Bioscience™ BS 181 dihydrochloride
Selective cdk7 inhibitor
Brand: Tocris Bioscience™ 5608/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
BS 181 dihydrochloride is a selective cdk7 inhibitor (IC50 = 21 nM). Displays >40-fold selectivity for cdk7 over others cdks. Promotes cell cycle arrest and apoptosis in a range of cancer cells in vitro and inhibits MCF-7 tumor xenograft growth in mice.Specifikationer
| N5-(6-Aminohexyl)-3-(1-methylethyl)-N7-(phenylmethyl)pyrazolo[1,5-a]pyrimidine-5,7-diamine dihydrochloride | |
| 1883548-83-1 | |
| 50 mg |