Läs mer
Tocris Bioscience™ BS 181 dihydrochloride
Beskrivning
BS 181 dihydrochloride is a selective cdk7 inhibitor (IC50 = 21 nM). Displays >40-fold selectivity for cdk7 over other cdks. Promotes cell cycle arrest and apoptosis in a range of cancer cells in vitro and inhibits MCF-7 tumor xenograft growth in mice.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N5-(6-Aminohexyl)-3-(1-methylethyl)-N7-(phenylmethyl)pyrazolo[1,5-a]pyrimidine-5,7-diamine dihydrochloride |
| CAS | 1883548-83-1 |
| Kvantitet | 10 mg |
| Mål | Cyclin-dependent Kinase Inhibitors |
| Molekylformel | C22H32N6.2HCl |
| Formel vikt | 453.45 |
| Renhet | >99% |