Läs mer
Paragon Scientific™ BS/IP/RF U-Tube Reverse Flow for Transparent and Opaque Liquids
Viskositetsområde:
0.6 to 3 cSt
2 to 10 cSt
20 to 100 cSt
20,000 to 100,000 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
60,000 to 300,000 cSt
600 to 3000 cSt
6000 to 30,000 cSt
Förpackningsstorlek:
Varje
Beskrivning
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifikationer
Specifikationer
| Viskositetsområde | 20,000 to 100,000 cSt |
| Kalibrerad | No |
| Ungefärlig konstant | 100 |
| ASTM-nummer | D 445, D 446 |
| Certifikat och efterlevnad | ASTM D 445, D 446 and ISO 3104, 3105 |
| Storlek | 10 |
4 week lead-time