Learn More
Paragon Scientific™ BS/IP/RF U-Tube Reverse Flow for Transparent and Opaque Liquids
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand:  Paragon Scientific™ RF08NC
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 16060330
  • 1507.00 SEK / Each
Estimated Shipment
22-03-2021
Add to basket
Description
Description
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
D 445, D 446
ASTM D 445, D 446 and ISO 3104, 3105
2000 to 10,000 cSt
10
8
Videos
SDS
Documents
4 week lead-time