Learn More
Paragon Scientific™ BS/IP/SL(S) Viscometer for Transparent Liquids
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand: Paragon Scientific™ SHORT05NC
Additional Details : Weight : 0.40000kg
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
20 to 100 cSt | |
0.1 | |
ASTM D 445, D 446 and ISO 3104, 3105 |
No | |
D 445, D 446 | |
5 |
4 week lead-time