missing translation for 'onlineSavingsMsg'
Learn More

BTBD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15859-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825244

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The C-terminus of BTBD1 binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications

Specifications

BTBD1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P58544, Q9H0C5
BTB (POZ) domain containing 1, BTBD1, C15orf1, NS5ATP8
Rabbit
IgG
150 μL
-20°C
Primary
293060, 53339, 83962
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BTBD1
Liquid
Btbd1
BTBD1 Fusion Protein Ag8339
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers