missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ BTBD2 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 15827053
Change view
Click to view available options
Kvantitet:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
15827053 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15827053 Supplier Invitrogen™ Supplier No. PA568639

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 90%.

BTBD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD2
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Formulering PBS with 2% sucrose and 0.09% sodium azide
Gen BTBD2
Genaccessionsnr. Q9BX70
Gene Alias 2610037C03Rik; 4930512K17Rik; BTB (POZ) domain containing 2; BTB domain containing 2; BTB/POZ domain-containing protein 2; BTBD2; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein 2; RGD1566094
Gensymboler BTBD2
Värd art Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human BTBD2.
Reningsmetod Affinity Chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 55643
Målarter Human
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.