missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN2A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20596-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The specific function of this protein remains unknown. This gene is a member of the BTN2 subfamily of genes, which encode proteins belonging to the butyrophilin protein family. The gene is located in a cluster on chromosome 6, consisting of seven genes belonging to the expanding B7/butyrophilin-like group, a subset of the immunoglobulin gene superfamily. The encoded protein is an integral plasma membrane B box protein involved in lipid, fatty-acid and sterol metabolism.Specifications
BTN2A1 | |
Polyclonal | |
Unconjugated | |
BTN2A1 | |
BK14H9.1, BT2.1, BTF1, BTN2A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11120 | |
-20°C | |
Liquid |
Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7KYR7 | |
BTN2A1 | |
BTN2A1 Fusion Protein Ag14545 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |