missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN3A3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | BTN3A3 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895284
|
Proteintech
15896-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16885284
|
Proteintech
15896-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Plays a role in T-cell responses in the adaptive immune response.Specifications
BTN3A3 | |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O00478 | |
10384 | |
BTN3A3 Fusion Protein Ag8711 | |
Primary | |
-20°C | |
BTN3A3 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BTF3, BTN3A3 | |
BTN3A3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |