missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN3A3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15896-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Plays a role in T-cell responses in the adaptive immune response.Specifications
BTN3A3 | |
Polyclonal | |
Unconjugated | |
BTN3A3 | |
BTF3, BTN3A3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10384 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00478 | |
BTN3A3 | |
BTN3A3 Fusion Protein Ag8711 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |