Läs mer
BTS 44595, TRC
Produktkod. 30138720
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138720 100 mg
17912885 50 mg
17944525 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138720 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB689350100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139520-94-8
Molekylformel C12 H15 Cl3 N2 O2
Molekylvikt (g/mol) 325.62
Synonym N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]urea, BTS 44595
IUPAC-namn 1-propyl-1-[2-(2,4,6-trichlorophenoxy)ethyl]urea
LEDER CCCN(CCOc1c(Cl)cc(Cl)cc1Cl)C(=O)N
Specifikationer
Kemiskt namn eller material Prochloraz desimidazole-amino
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H15Cl3N2O2/c1-2-3-17(12(16)18)4-5-19-11-9(14)6-8(13)7-10(11)15/h6-7H,2-5H2,1H3,(H2,16,18)
Formel vikt 324.0199
Procent renhet >95%