Learn More
BTX™ HT96 Enhancer™ 3000 Electroporation/Monitoring Systems
Versatile systems combine rapid multi-well electroporation with customizable data monitoring and analysis for fast, accurate results
POA
Specifications
For Use With (Application) | Transformation of bacteria and yeast, transfection of mammalian cells |
---|---|
Includes | ECM generator, HT-200 plate handler, two 96-well HT plates, enhancer 3000 monitoring system with communications module |
Includes:
- ECM Generator
- HT-200 Plate Handler
- Two 96-well HT plates
- Enhancer 3000 Monitoring System with communications module
Description
The HT-96 Multi-Well System uses 96-well plates for faster turnaround than traditional cuvets. Run multiple experiments simultaneously, or load each plate with the same sample for high throughput processing. The Enhancer 3000 System enables the user to observe waveform, confirm data, track results, and document key parameters for protocol optimization, troubleshooting, and routine quality control.
- Processes multiple samples in seconds for optimization of electroporation variables
- Complete user control over electroporation settings allows for a wide range of optimization flexibility
- Results are confirmed automatically
- Monitoring and documentation of waveforms and electroporation parameters lets you quickly determine potential problems
- Communications module offers standard printer and computer interface
Choice of Generators
- ECM 630, an exponential decay generator recommended for high throughput electroporation of bacterial and yeast samples
- ECM 830, a square wave generator, for biomolecular screening, transfection, and gene silencing with no cytotoxicity
Specifications
Transformation of bacteria and yeast, transfection of mammalian cells | |
ECM generator, HT-200 plate handler, two 96-well HT plates, enhancer 3000 monitoring system with communications module |