Learn More
BTX™ HT96 Enhancer™ 3000 Electroporation/Monitoring Systems
Versatile systems combine rapid multi-well electroporation with customizable data monitoring and analysis for fast, accurate results
Brand:  BTX Molecular Delivery Systems 450472
Additional Details : Weight : 12.90000kg
 View more versions of this product
Product Code. 15477320
  • 193285.00 SEK / 1 set
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Specifications
Specifications
ECM generator, HT-200 plate handler, two 96-well HT plates, enhancer 3000 monitoring system with communications module
Transformation of bacteria and yeast, transfection of mammalian cells
ECM 830 generator