Learn More
BUCHI Glass Stopcock for Rotavapor condensers
For aeration of Rotavapor systems or sample addition depending on configuration
Brand:  BUCHI 040627
Additional Details : Weight : 0.07200kg
 View more versions of this product
Product Code. 10017061
  • 845.00 SEK / Each
Estimated Shipment: 28-08-2024
to see stock.
Add to basket
Description
Description
  • OEM BUCHI replacement
  • Precision ground joint for reliable sealing and operation
Specifications
Specifications
Stopcock
Rotavapor model R-200
SJ18.8/38
18.8/38