Learn More
BUCHI Set of Bent and Straight Hose Barbs with GL14 threads for condensers
Package contains hose barbs, cap nuts, and seals for BUCHI Rotavapor&trade vacuum and chiller connections.
Brand:  BUCHI 038000
Additional Details : Weight : 0.03000kg
Product Code. 12005492
  • 768.49 SEK / 6 sets
Estimated Shipment: 07-06-2022
to see stock.
Add to basket
Description
Description
  • Hose barb set, 6 pcs
  • Bent (4), straight (2)
  • GL14 caps
  • Silicone seals
Specifications
Specifications
Hose Barb Set
Vacuum and chiller connections
Polypropylene
GL14
Hose Barb
8 mm
Bent (4), Straight (2)
30 mm