missing translation for 'onlineSavingsMsg'
Learn More

BUD13 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20163-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16879664

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BUD13 is a protein coding gene involved in pre-mRNA splicing as component of the activated spliceosome.
Specifications

Specifications

BUD13
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BRD0
BUD13, BUD13 homolog, BUD13 homolog (S. cerevisiae), fSAP71
Rabbit
IgG
20 μL
-20°C
Primary
84811
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
BUD13
Liquid
BUD13
BUD13 Fusion Protein Ag13665
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers