missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUD13 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | BUD13 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879664
|
Proteintech
20163-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16869664
|
Proteintech
20163-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
BUD13 is a protein coding gene involved in pre-mRNA splicing as component of the activated spliceosome.Specifications
BUD13 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9BRD0 | |
84811 | |
BUD13 Fusion Protein Ag13665 | |
Primary | |
-20°C | |
BUD13 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BUD13, BUD13 homolog, BUD13 homolog (S. cerevisiae), fSAP71 | |
BUD13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |