Learn More
Buffer solution pH 10.00 (+/-0.024 @ 25oC) Colored Blue Specpure NIST Traceable, Thermo Scientific Chemicals
569.20 SEK - 1048.47 SEK
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398977
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
038709.K2
1 L
569.20 SEK
1 L
View alternative products
Save Up to 
»
In Stock
to see stock.
11308987
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
038709.K7
4 L
1048.47 SEK
4 L
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Description
Description
Buffer solution (pH 10.00) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.