missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10.00, NIST, Chem-Lab
Click to view available options
Quantity:
10 L
25 L
Description
pH = 10.24 at 0°C
pH = 10.10 at 10°C
pH = 10.00 at 20°C
pH = 9.95 at 25°C
pH = 9.90 at 30°C
pH = 9.82 at 40°C
pH = 9.75 at 50°C
pH = 9.68 at 60°C
pH = 9.62 at 70°C
pH = 9.55 at 80°C
pH = 9.49 at 90°C
pH = 10.10 at 10°C
pH = 10.00 at 20°C
pH = 9.95 at 25°C
pH = 9.90 at 30°C
pH = 9.82 at 40°C
pH = 9.75 at 50°C
pH = 9.68 at 60°C
pH = 9.62 at 70°C
pH = 9.55 at 80°C
pH = 9.49 at 90°C
Specifications
Density | 1.01g/mL |
pH | 10.0 |
Quantity | 25 L |
Formulation | H3BO3, KCl, NaOH |
Packaging | PE |
Traceability to NIST | NIST traceable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'