missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00, NIST, Chem-Lab

Catalog Number p-7114730
Click to view available options
Quantity:
10 L
25 L
This item is not returnable. View return policy
This item is not returnable. View return policy

For laboratory use, pH measurements, traceable to SRM from NIST

pH = 10.24 at 0°C
pH = 10.10 at 10°C
pH = 10.00 at 20°C
pH = 9.95 at 25°C
pH = 9.90 at 30°C
pH = 9.82 at 40°C
pH = 9.75 at 50°C
pH = 9.68 at 60°C
pH = 9.62 at 70°C
pH = 9.55 at 80°C
pH = 9.49 at 90°C
Density 1.01g/mL
pH 10.0
Quantity 10 L
Formulation H3BO3, KCl, NaOH
Packaging PE
Traceability to NIST NIST traceable
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'