Läs mer
Buflomedil Hydrochloride, TRC
Produktkod. 30348577
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30348577 10 g
1 options
Produktkod. 30348577 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68940010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Buflomedil hydrochloride
CAS 35543-24-9
Molekylformel C17 H25 N O4 . Cl H
Kvantitet 10 g
Synonym Buflomedil Hydrochloride, 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone hydrochloride, Bufedil, Buflan, Buflocit, Buflomedil hydrochloride, Buflonat, Fonzylane, Irrodan, Irrodan R, LL 1656, Lofton, Loftyl, Provas
InChI formel InChI=1S/C17H25NO4.ClH/c1-20-13-11-15(21-2)17(16(12-13)22-3)14(19)7-6-10-18-8-4-5-9-18;/h11-12H,4-10H2,1-3H3;1H
LEDER Cl.COc1cc(OC)c(C(=O)CCCN2CCCC2)c(OC)c1
IUPAC-namn 4-pyrrolidin-1-yl-1-(2,4,6-trimethoxyphenyl)butan-1-one;hydrochloride
Molekylvikt (g/mol) 343.85
Formel vikt 343.155