Läs mer
Bufuralol-d9 Hydrochloride, TRC
Produktkod. 30330649
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30330649 1 mg
30347741 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30330649 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB6895421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Bufuralol-d9 Hydrochloride
CAS 1173023-51-2
Molekylformel C16H15D9ClNO2
Kvantitet 1 mg
Synonym α-[[(1,1-Dimethylethyl)-amino]methyl]-7-ethyl-2-benzofuranmethanol-d9, Ro-3-4787-d9, Angium-d9
InChI formel InChI=1S/C16H23NO2.ClH/c1-5-11-7-6-8-12-9-14(19-15(11)12)13(18)10-17-16(2,3)4;/h6-9,13,17-18H,5,10H2,1-4H3;1H/i2D3,3D3,4D3;
LEDER OC(CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C(O1)=CC2=C1C(CC)=CC=C2.Cl
IUPAC-namn 1-(7-ethyl-1-benzofuran-2-yl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]ethanol;hydrochloride
Molekylvikt (g/mol) 306.88